Robotic tips (CO-RE Tips) and consumables for Microlab
Product Details & Documents
Hamilton CO-RE tips were designed for MICROLAB pipetting workstations in order to solve the problem of sealing and alignment plagued by press-fit tip attachment techniques. Each CO-RE disposable tip has a circular groove inside the attachment orifice. An o-ring on the pipetting channel is expanded to fit this groove precisely providing an air-tight seal and straight alignment between tip and pipetting channel.
- Pressure-based Liquid Level Detection (pLLD) and capacitive LLD (cLLD) with conductive (black) version
- DNase-/RNase-free, pyrogen-free (endotoxin-free), PCR inhibitor-free
- Stackable tip racks increase tip capacity on the workstation deck for longer walk away times: 1920 tips can be loaded in one step
- Racks allow tips to be picked up by the independent channels or the 96-channel multiprobe head directly from the stack
The superior biological purity of the tips is guaranteed by a controlled production process. They are therefore suitable for both diagnostic and pharmaceutical laboratories in Genomics, Proteomics, Cellomics, Forensic and drug discovery fields.
Filter tips
- Maximum consistency in air flow and chosen porosity of filter element ensure optimal pipetting and dispensing performance across a wide range of liquid volumes
- Bacterial aerosol filtration efficiency of >99,9%
SLIM tips
- Minimise risk of liquid overflow in cluster tubes, deep-well blocks and two-phased liquid applications
- Especially for aspirating liquid in 'fixed height mode' down to the base of thin tubes
- Slim tips require labware files, liquid classes for single channels and CO-RE 96 probe head
Rocket tips
- 4-to-1 adapter enables flexible pipetting and saves times: No need for a second head or a complex head exchange
- For pipetting in 96 deep-well blocks and microtiter plates with well diameters ≥3,7 mm
Wide bore tips
- CO-RE tips with various distal end orifices which provide the flexibility when handling difficult samples
- Typical application areas: genomic DNA, fragile cell lines, blood fractionation to minimise shearing forces, pipetting beads or other viscosity liquids
Piercing tips
- Optimised to aspirate and dispense into various sealed labware which is pierceable with ≤10 N
- Optimum material composition – carbon fibre reinforcement allows piercing of various sealing materials
SLIM tips
- Minimise risk of liquid overflow in cluster tubes, deep-well blocks and two-phased liquid applications
- Especially for aspirating liquid in “fixed height mode” down to the base of thin tubes
- 40% reduction of displaced liquid volume, compared to the CO-RE 1000 µl tip
- Slim tips require labware files, liquid classes for single channels and CO-RE 96 probe head
Rocket tips
- Unique Rocket tip design with 4-to-1 adapter: Usable on the CO-RE 384 probe head
- Adapter enables flexible pipetting and saves times: No need for a second head or a complex head exchange
- For pipetting in 96 deep-well blocks and microtiter plates with well diameters ≥ 3,7 mm
Zubehördetails: Reagent containers for Microlab STAR line
- Made of PP, autoclavable, with 60 or 120 ml capacity
- Can be placed in a reagent carrier on STAR line instruments
- With volume markings, without lid, with one (60 ml) or three (120 ml) wave breakers to avoid splashes during movements of the filled container
Deep-well blocks for automation
- Ideal for automated processing or handling large quantities of backup samples
- PP material is ideal for backup storage of pharmaceutical substances, DNA isolation, long-term sample storage, enzyme and tissue culture applications
- PS blocks are ideal for long-term storage of blood samples; easy optical control due to transparent wells
Waste bags, liquid-tight, welded
- Disposable PE bags (100 µm thick) are ideal for the containment and disposal of used consumables
- Bags fit on the waste hoop of the STAR line
Waste container and chutes for hazardous waste
- Container for hazardous waste is made of white cardboard, incl. yellow PE bag
- PE waste chutes are open and perforated on both sides, must be used with waste container
Verpackung: Tips are either supplied in stackable Nested Tip Rack (NTR) packs – containing a disposable frame and five stacks with four tip racks – or in a blister pack containing five preloaded trays in a clear, recyclable PET thermoformed tray with a sealed paper lid.
Spezifikationen:
- Packed:24 × 1T tray (each 4 tips individually wrapped)
- Sterile:-
- Volume:50 - 5000 µl
- Length:118 mm
- Seq:40
- Tip style:High volume, conductive
- Tip colour:Black
- Filter:-
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



