ContactUsLinkComponent
pH combination electrode with Memosens screw plug head Liquid electrolyt comb. electrode with NTC 30K and plastic adapter for hose c onnection incl. Pg13.5 thread, platinum junction, electrolyte L 200, sphere membrane, A-glass, length 380 mm, 12 mm Ø 1 * 1 items
Catalog # SCOI285118195
Supplier:  SI Analytics
pH combination electrode with Memosens screw plug head Liquid electrolyt comb. electrode with NTC 30K and plastic adapter for hose c onnection incl. Pg13.5 thread, platinum junction, electrolyte L 200, sphere membrane, A-glass, length 380 mm, 12 mm Ø 1 * 1 items
Catalog # SCOI285118195
Supplier:  SI Analytics
Frequently Bought Together