ContactUsLinkComponent
ACQUITY UPLC HSS C18 SB Method Validation Kit, 100Å, 1.8µm, 2.1mm x 50mm (Kit consists of three ACQUITY UPLC HSS C18 SB columns packed with three different batches of sorbent) 1 * 1 KIT
Catalog # WATE186004137
Supplier:  WATERS
ACQUITY UPLC HSS C18 SB Method Validation Kit, 100Å, 1.8µm, 2.1mm x 50mm (Kit consists of three ACQUITY UPLC HSS C18 SB columns packed with three different batches of sorbent) 1 * 1 KIT
Catalog # WATE186004137
Supplier:  WATERS