Venipuncture sets, Surflo®
Winged needles with protector, G 21, Green, 19 mm
TERUSV-S21NL30
: TERUMO
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.
- Gauge:G 21
- Colour:Green
- Description:Winged needles with protector
- Length:19 mm
- Seq:002