Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Osmolality standards

Osmolality standard, 850 mOsm/kg H₂O, Ampoule, 12×5 ml, 1 KIT
Catalog # REGERE-OSM-850
Supplier:   REAGECON
Osmolality standards
Catalog # REGERE-OSM-850
Supplier:  REAGECON
CAS Number:  
Osmolality standards

Osmolality standard, 850 mOsm/kg H₂O, Ampoule, 12×5 ml, 1 KIT

CAS Number:  
Price
183,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.

  • Ready to use and presented in convenient ampoules
  • Extended shelf life and has low uncertainty of measurement
  • Consistency of product-Independent, traceable
  • Consist of 12 ampoules of 5 ml

The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.

Certifications: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.

Documents

Specifications

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1 KIT
  • Description:
    Osmolality standard
  • Pk Info:
    12×5 ml
  • Osmolality:
    850 mOsm/kg H₂O

Specifications