Order Entry
Germany
Orders LinkContactUsLinkComponent
 

EasySep™ Red Blood Cell Lysis Buffer

Red blood cell lysis buffer, 10X concentrate, 100 mL
  STMC20120
Marketsource
EasySep™ Red Blood Cell Lysis Buffer
  STMC20120
Marketsource
 :  
EasySep™ Red Blood Cell Lysis Buffer
EasySep™ Red Blood Cell Lysis Buffer

Red blood cell lysis buffer, 10X concentrate, 100 mL

 :  
 
695,00 €
   
1 * 100 mL
 

EasySep™ Red blood cell lysis buffer is for use as directed with EasySep™ or RoboSep™ whole blood kits for the lysis of red blood cells prior to separation from whole blood.

Application: Chimerism, Hla, immunology
Species: Human

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

 

 

  • Pk:
    100 mL
  • Description:
    Red blood cell lysis buffer, 10X concentrate

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.