Order Entry
Germany
Orders LinkContactUsLinkComponent
Process pH combination electrodes

Process pH combination electrodes

pH combination electrode, with cylinder membrane and PT1000, H 7381-T1000 VP, 2 - 13, −5...+80 °C, Glass, Gel, Ceramic, 12×120 mm, VP plug head
Catalog # SCOI285090155
Supplier:   SI Analytics
Process pH combination electrodes
Catalog # SCOI285090155
Supplier:  SI Analytics
Process pH combination electrodes
Process pH combination electrodes

pH combination electrode, with cylinder membrane and PT1000, H 7381-T1000 VP, 2 - 13, −5...+80 °C, Glass, Gel, Ceramic, 12×120 mm, VP plug head

Price
387,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Connection:
    VP plug head
  • Ø×L:
    12×120 mm
  • Type:
    H 7381-T1000 VP
  • Description:
    pH combination electrode, with cylinder membrane and PT1000
  • Material:
    Glass
  • Seq:
    11
  • Diaphr.:
    Ceramic
  • Electrolyte:
    Gel
  • Temp.:
    −5...+80 °C
  • pH:
    2 - 13