Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Mob caps, PP

Red, 520 mm
  113-9197
Mob caps, PP
  113-9197
Mob caps, PP
Mob caps, PP

Red, 520 mm

 
 
   
1 * 1.000 ST
 

Disposable mob caps made of PP. Ideal for use in non critical environments.

  • With single latex-free elastic band
  • Available in different colours

Weight: 10 to 12 g/m²

 : 100 pieces per polybag bag and 1000 pieces per carton.

 
  • Ø:
    520 mm
  • Colour:
    Red
  • Seq:
    04
  • Cat. No.:
    113-9197
  • Garment application:
    General Purpose