Order Entry
Germany
Orders LinkContactUsLinkComponent
 

5'-Bromo-2'-hydroxyacetophenone ≥98%

Glass bottle for solids, 50 g
Catalog # H61979.18
Irritant
5'-Bromo-2'-hydroxyacetophenone ≥98%
Catalog # H61979.18
CAS Number:  1450-75-5
5'-Bromo-2'-hydroxyacetophenone ≥98%
5'-Bromo-2'-hydroxyacetophenone ≥98%

Glass bottle for solids, 50 g

CAS Number:  1450-75-5
Price
224,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
  • CAS Number:1450-75-5
  • Formula:C₈H₇BrO₂
  • MDL Number:MFCD00191850
  • Molecular Weight:215.05
  • Melting Pt:56…60 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications