ContactUsLinkComponent
LOC554223 (Human) Recombinant protein (P01). Human LOC554223 full-length ORF ( AAH51362.1, 1 A.A. - 189 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00554223-P012
Supplier:  Abnova
LOC554223 (Human) Recombinant protein (P01). Human LOC554223 full-length ORF ( AAH51362.1, 1 A.A. - 189 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00554223-P012
Supplier:  Abnova
Frequently Bought Together