ContactUsLinkComponent
LOC493829 (Human) Recombinant protein (P01). Human LOC493829 full-length ORF ( AAH33211.1, 1 A.A. - 269 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00493829-P012
Supplier:  Abnova
LOC493829 (Human) Recombinant protein (P01). Human LOC493829 full-length ORF ( AAH33211.1, 1 A.A. - 269 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00493829-P012
Supplier:  Abnova
Frequently Bought Together