ContactUsLinkComponent
LOC155060 (Human) Recombinant protein (P01). Human LOC155060 full-length ORF ( AAI31570.1, 1 A.A. - 281 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00155060-P012
Supplier:  Abnova
LOC155060 (Human) Recombinant protein (P01). Human LOC155060 full-length ORF ( AAI31570.1, 1 A.A. - 281 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00155060-P012
Supplier:  Abnova
Frequently Bought Together