ContactUsLinkComponent
Human PCNP full-length ORF ( NP_065090.1, 1 A.A. - 178 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 G
Catalog # ABNOH00057092-P012
Supplier:  Abnova
Human PCNP full-length ORF ( NP_065090.1, 1 A.A. - 178 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 G
Catalog # ABNOH00057092-P012
Supplier:  Abnova