ContactUsLinkComponent
KIAA0804 (Human) Recombinant protein (Q01). Human KIAA0804 partial ORF ( NP_001009921, 1329 A.A. - 1428 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00023355-Q012
Supplier:  Abnova
KIAA0804 (Human) Recombinant protein (Q01). Human KIAA0804 partial ORF ( NP_001009921, 1329 A.A. - 1428 A.A.) Recombinant protein wi th GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00023355-Q012
Supplier:  Abnova
Frequently Bought Together