Order Entry
Germany
ContactUsLinkComponent
Calibration gas 50 h2s,200 co,2.5% ch4,17% o2,n2 34l 4ga17x25w50h200-34 1 * 1 items
Catalog # BWTE4GA17X25W50H20
Supplier:  BW TECHNOLOGIES
Calibration gas 50 h2s,200 co,2.5% ch4,17% o2,n2 34l 4ga17x25w50h200-34 1 * 1 items
Catalog # BWTE4GA17X25W50H20
Supplier:  BW TECHNOLOGIES