ContactUsLinkComponent
Human ACE partial ORF ( AAH36375, 592 A.A. - 701 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00001636-Q012
Supplier:  Abnova
Human ACE partial ORF ( AAH36375, 592 A.A. - 701 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00001636-Q012
Supplier:  Abnova