Orders LinkContactUsLinkComponent
Boxes for pipette tips
Boxes for pipette tips
Empty TipBox, PP, with tip-tray, stackable, for tips up to 1000 µl, Blue
Catalog # 613-2427
Supplier:   Brand
Boxes for pipette tips
Catalog # 613-2427
Supplier:  Brand
Boxes for pipette tips
Boxes for pipette tips
Empty TipBox, PP, with tip-tray, stackable, for tips up to 1000 µl, Blue
Price
5,55 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Colour code:
    Blue
  • Description:
    Empty TipBox, PP, with tip-tray, stackable, for tips up to 1000 µl
  • Seq:
    04