ContactUsLinkComponent
standard interior equipment gas cylinders consisting of: mounting rails, cylinder retainer and rolling ramp, sheet steel, galvanized and plastic laminated for tall cabinets with width 600 mm for model(s): g90 1 * 1 items
Catalog # MSGA30782
Supplier:  asecos
standard interior equipment gas cylinders consisting of: mounting rails, cylinder retainer and rolling ramp, sheet steel, galvanized and plastic laminated for tall cabinets with width 600 mm for model(s): g90 1 * 1 items
Catalog # MSGA30782
Supplier:  asecos
Frequently Bought Together