Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillary, pre-swaged (from valve with 10/32 Swagelok port to heat exchanger), 0,28 m, I.Ø 0,12 mm
Catalog # HEWL5067-4649
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWL5067-4649
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems
Capillary, pre-swaged (from valve with 10/32 Swagelok port to heat exchanger), 0,28 m, I.Ø 0,12 mm
Price
96,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Description:
    Capillary, pre-swaged (from valve with 10/32 Swagelok port to heat exchanger), 0,28 m, I.Ø 0,12 mm
  • Seq:
    81