SONOREX DIGITEC DT 100, ultrasonic bath, 230 V~ 50/60 Hz, ID 240x140x100 mm 3,0 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15,30 min and continuous, ultrasonic peak output 320 W, HF output 80 Weff., SweepTec and DEGAS 1 * 1 items
Catalog # BAND3210
Supplier: BANDELIN-ELECTRONIC