Capillaries for LC systems
Capillary, non-swaged (from autosampler to valve with M4 port), 0,50 m, I.Ø 0,11 mm
HEWL5067-4745
: Agilent
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Description:Capillary, non-swaged (from autosampler to valve with M4 port), 0,50 m, I.Ø 0,11 mm
- Seq:73