Order Entry
Germany
Orders LinkContactUsLinkComponent
Superdex 200 Increase small-scale SEC columns

Superdex 200 Increase small-scale SEC columns

Superdex™ 200 Increase 5/150 GL, Glass, 5×150 mm, 3 ml, 0,004 - 0,050 ml, <0,75 ml/min
Catalog # 28-9909-45
Supplier:   Cytiva
Superdex 200 Increase small-scale SEC columns
Catalog # 28-9909-45
Supplier:  Cytiva
Superdex 200 Increase small-scale SEC columns
Superdex 200 Increase small-scale SEC columns

Superdex™ 200 Increase 5/150 GL, Glass, 5×150 mm, 3 ml, 0,004 - 0,050 ml, <0,75 ml/min

Price
2 960,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Superdex 200 Increase size exclusion chromatography columns are well suited for high-resolution analysis and small-scale purification of monoclonal antibodies (mAb) and other biomolecules with Mr~ 10000 to 600000.

  • Enhanced performance: Improved resolution and runtime compared to predecessor Superdex 200
  • Very high resolution: Small bead size and narrow particle size distribution provide high resolution, for high protein purity
  • High flow rates: Rigid beads give excellent pressure/flow properties

Superdex 200 Increase is a next-generation, dextran-agarose composite matrix for SEC. With smaller and more rigid beads than their predecessors, Superdex Increase columns deliver higher resolution purification in shorter run times.

Specifications
  • Flow rate:
    <0,75 ml/min
  • Capacity:
    3 ml
  • Description:
    Superdex™ 200 Increase 5/150 GL
  • Seq:
    02
  • Column hardware:
    Glass
  • Bed dimensions:
    5×150 mm
  • Sample volume:
    0,004 - 0,050 ml
  • Cat. No.:
    28-9909-45
  • Max. pressure:
    30 bar
  • Storage:
    4 to 30 °C, 20% Ethanol
  • pH range:
    3 - 12