Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solution for chlorine determination

Buffer solution for chlorine determination

7, Phosphate, 1
Catalog # HACH2155332
Supplier:   Hach
Buffer solution for chlorine determination
Catalog # HACH2155332
Supplier:  Hach
CAS Number:  
Buffer solution for chlorine determination
Buffer solution for chlorine determination

7, Phosphate, 1

CAS Number:  
Price
29,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

For determination of chlorine by amperometric titration. APHA formulation 100 ml bottle.

Specifications

Specifications

  • pH buffer type:
    Phosphate
  • Pk:
    1
  • pH value:
    7

Specifications