ContactUsLinkComponent
The hydrogen-specific triple trap is ideal for purifying hydrogen in gc/ ms systems. this trap is packed and purged under hydrogen a nd contains oxygen, moisture and hydrocarbon scrubbers in one cartridge. its features and benefits include - ultra-hi 1 * 1 items
Catalog # PERCN9306822
Supplier:  PerkinElmer
The hydrogen-specific triple trap is ideal for purifying hydrogen in gc/ ms systems. this trap is packed and purged under hydrogen a nd contains oxygen, moisture and hydrocarbon scrubbers in one cartridge. its features and benefits include - ultra-hi 1 * 1 items
Catalog # PERCN9306822
Supplier:  PerkinElmer
Frequently Bought Together