ContactUsLinkComponent
Moisture in carrier gas lines will prematurely degrade oxygen and hydrocarbon traps and increase detector noise. as a precaution, we higly recommend installing a moisture trap before the hydrocarbon and oxygen traps on all carrier gas lines. its fea 1 * 1 items
Catalog # PERCN9306814
Supplier:  PerkinElmer
Moisture in carrier gas lines will prematurely degrade oxygen and hydrocarbon traps and increase detector noise. as a precaution, we higly recommend installing a moisture trap before the hydrocarbon and oxygen traps on all carrier gas lines. its fea 1 * 1 items
Catalog # PERCN9306814
Supplier:  PerkinElmer