ContactUsLinkComponent
Triple position ultra clean filter base plate for gc. a base plate allows cartridges to be exchanged without introducing oxygen. its features and benefits include - ultra-high capacity high-purity output ensures 99.9999% pure gas quick connect fit 1 * 1 items
Catalog # PERCN9306811
Supplier:  PerkinElmer
Triple position ultra clean filter base plate for gc. a base plate allows cartridges to be exchanged without introducing oxygen. its features and benefits include - ultra-high capacity high-purity output ensures 99.9999% pure gas quick connect fit 1 * 1 items
Catalog # PERCN9306811
Supplier:  PerkinElmer
Frequently Bought Together