ContactUsLinkComponent
drawer disposal, sheet steel, plastic laminated plain, ral 7035 light grey load capacity (evenly distributed load) (kg): 50 volume ( l): 30 for medium and tall cabinets with width 600 mm for model(s): q90, s90, s60 1 * 1 items
  MSGA30109
 :  asecos
drawer disposal, sheet steel, plastic laminated plain, ral 7035 light grey load capacity (evenly distributed load) (kg): 50 volume ( l): 30 for medium and tall cabinets with width 600 mm for model(s): q90, s90, s60 1 * 1 items
  MSGA30109
 :  asecos
Frequently Bought Together