ContactUsLinkComponent
drawer heavy duty, stainless steel, raw load capacity (evenly distributed load) (kg): 100 volume (l): 45 for tall cabinets with widt h 900 mm for model(s): q90, s90 1 * 1 items
  MSGA30068
 :  asecos
drawer heavy duty, stainless steel, raw load capacity (evenly distributed load) (kg): 100 volume (l): 45 for tall cabinets with widt h 900 mm for model(s): q90, s90 1 * 1 items
  MSGA30068
 :  asecos
Frequently Bought Together