ContactUsLinkComponent
pull-out shelf with sump, sheet steel/polyethylene, plastic laminated/raw, ral 7035 light grey load capacity (evenly distributed loa d) (kg): 30 volume (l): 25 for medium and tall cabinets with width 810 mm for model(s): cs 1 * 1 items
  MSGA30556
 :  asecos
pull-out shelf with sump, sheet steel/polyethylene, plastic laminated/raw, ral 7035 light grey load capacity (evenly distributed loa d) (kg): 30 volume (l): 25 for medium and tall cabinets with width 810 mm for model(s): cs 1 * 1 items
  MSGA30556
 :  asecos