ContactUsLinkComponent
shelf standard, sheet steel, galvanized load capacity (evenly distributed load) (kg): 50 for tall cabinets with width 950 mm for mod el(s): em 1 * 1 items
Catalog # MSGA30640
Lieferant:  asecos
shelf standard, sheet steel, galvanized load capacity (evenly distributed load) (kg): 50 for tall cabinets with width 950 mm for mod el(s): em 1 * 1 items
Catalog # MSGA30640
Lieferant:  asecos
Frequently Bought Together