ContactUsLinkComponent
recombinant protein, fkbp6 (human) recombinant protein (p01), human fkbp6 full-length orf ( np_001013770.1, 1 a.a. - 131 a.a.) recom binant protein with gst-tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00541473-P01
Supplier:  Abnova
recombinant protein, fkbp6 (human) recombinant protein (p01), human fkbp6 full-length orf ( np_001013770.1, 1 a.a. - 131 a.a.) recom binant protein with gst-tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00541473-P01
Supplier:  Abnova