ContactUsLinkComponent
recombinant protein, sp3 (human) recombinant protein (q02), human sp3 partial orf (np_003102.1, 516 a.a. - 615 a.a.) recombinant pro tein with gst tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00006670-Q02
Supplier:  Abnova
recombinant protein, sp3 (human) recombinant protein (q02), human sp3 partial orf (np_003102.1, 516 a.a. - 615 a.a.) recombinant pro tein with gst tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00006670-Q02
Supplier:  Abnova