ContactUsLinkComponent
recombinant protein, cnr2 (human) recombinant protein (q01), human cnr2 partial orf (np_001832.1, 302 a.a. - 360 a.a.) recombinant p rotein with gst tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00001269-Q01
Supplier:  Abnova
recombinant protein, cnr2 (human) recombinant protein (q01), human cnr2 partial orf (np_001832.1, 302 a.a. - 360 a.a.) recombinant p rotein with gst tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00001269-Q01
Supplier:  Abnova