Orders LinkContactUsLinkComponent
Inserts
Inserts
A 12/1 Insert for the gentle cleaning of delicate items by interrupting the spray jet, with perforations 7×7 mm (224×429×60 mm), G 7804, G 7883, G 7893, G 7883 CD, G 7835 CD, G 7836 CD, 234×459×221 mm
Catalog # 141-0335
Supplier:   MIELE
Inserts
Catalog # 141-0335
Supplier:  MIELE
Inserts
Inserts
A 12/1 Insert for the gentle cleaning of delicate items by interrupting the spray jet, with perforations 7×7 mm (224×429×60 mm), G 7804, G 7883, G 7893, G 7883 CD, G 7835 CD, G 7836 CD, 234×459×221 mm
Price
85,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    A 12/1 Insert for the gentle cleaning of delicate items by interrupting the spray jet, with perforations 7×7 mm (224×429×60 mm)
  • Seq:
    4
  • W×D×H:
    234×459×221 mm
  • For:
    G 7804, G 7883, G 7893, G 7883 CD, G 7835 CD, G 7836 CD