Order Entry
Germany
Orders LinkContactUsLinkComponent
PBS buffer

PBS buffer

PBS-P+ buffer 10X, includes 0,2 M phosphate buffer with 27 mM KCl, 1,37 M NaCl and 0,5% surfactant P20 (tween 20)., 1.000 mL
Catalog # 28-9950-84
Supplier:   Cytiva
PBS buffer
Catalog # 28-9950-84
Supplier:  Cytiva
CAS Number:  
PBS buffer
PBS buffer

PBS-P+ buffer 10X, includes 0,2 M phosphate buffer with 27 mM KCl, 1,37 M NaCl and 0,5% surfactant P20 (tween 20)., 1.000 mL

CAS Number:  
Price
206,00 €
Log in for availability
1 * 1.000 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Phosphate buffered saline solution, available with or without detergent, recommended as running buffer for small molecule analysis in Biacore systems. Concentrated stock solution, yielding pH 7,4 when diluted 10X.

  • Can be stored at room temperature, precipitation can occur at low temperature
  • Supplemented with DMSO
Specifications

Specifications

  • Pk:
    1.000 mL
  • Description:
    PBS-P+ buffer 10X, includes 0,2 M phosphate buffer with 27 mM KCl, 1,37 M NaCl and 0,5% surfactant P20 (tween 20).
  • Cat. No.:
    28-9950-84
  • For use with:
    Interaction analyses in Biacore systems, most commonly small molecule assays

Specifications

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.