Buffers, carbonate, 0,5 M solutions
Carbonate buffer solution, 0,5 M, 9,5, Glasflasche, 250 mL
Supplier: Thermo Fisher Scientific
Buffers, carbonate, 0,5 M solutions
Catalog # J63658.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 497-19-8
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00003494
Documents
Specifications
Specifications
- Konzentration:0,5 M
- Packung:Glasflasche
- VE:250 mL
- Bezeichnung:Carbonate buffer solution
- pH-Wert:9,5
- Cat. No.:J63658.AK
- CAS Nr.:497-19-8
- Odour/scent:Odourless
- Physical form:Liquid
- MDL No.:MFCD00003494
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



