Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 250 ml
Catalog # J63560.AK
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Catalog # J63560.AK
Supplier:  
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 250 ml

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Sodium acetate, 3M aq. soln., pH 5.2, autoclaved 1 * 125 mL Catalog #J63560.XE

Product J63560.XE is a replacement for J63560.AK but varies by Brand, Packaging or Quantity.

Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.