Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 125 mL
Catalog # J63560.XE
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Catalog # J63560.XE
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 125 mL

CAS Number:  
Price
61,90 €
Log in for availability
1 * 125 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.