Order Entry
Germany
Orders LinkContactUsLinkComponent
Stacking buffer (4X)

Stacking buffer (4X)

Stacking Buffer, 4X, 1 L
Catalog # J63450.K2
Stacking buffer (4X)
Catalog # J63450.K2
CAS Number:  
Stacking buffer (4X)
Stacking buffer (4X)

Stacking Buffer, 4X, 1 L

CAS Number:  
Price
108,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Physical Form: Liquid

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Stacking Buffer, 4X
  • Cat. No.:
    J63450.K2
  • Physical form:
    Liquid

Specifications