Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Phosphate, 0,5M buffer solution, pH 7,5

Phosphate buffer soln, 0,5 M, 7,5, 500 mL
  J63349.AP
Phosphate, 0,5M buffer solution, pH 7,5
  J63349.AP
 :  
Phosphate, 0,5M buffer solution, pH 7,5
Phosphate, 0,5M buffer solution, pH 7,5

Phosphate buffer soln, 0,5 M, 7,5, 500 mL

 :  
 
92,40 €
   
1 * 500 mL
 

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

 

 

  • Concentration:
    0,5 M
  • Pk:
    500 mL
  • Description:
    Phosphate buffer soln
  • pH value:
    7,5
  • Cat. No.:
    J63349.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00003527
  • CAS No.:
    7558-80-7