Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 4 L
Catalog # J63931.K7
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # J63931.K7
CAS Number:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 4 L

CAS Number:  
Price
419,00 €
Log in for availability
1 * 4 L
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236357

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)
  • Cat. No.:
    J63931.K7
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00236357
  • Appearance:
    Colourless

Specifications