Neutral red
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 553-24-2 Molecular Formula: C15H17ClN4 Formula Weight: 288.78 Physical Form: Powder Appearance: Dark green to black Melting Point: 290° dec. Beilstein Reference: 3918740 Merck Reference: 14,6488 MDL No.: MFCD00012651
Nuclear counterstain useful in pH range 6.8 - 8.0
Synonyms: Basisrot 5, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid
- CAS Number:553-24-2
- Formula:C₁₅H₁₇ClN₄
- MDL Number:MFCD00012651
- Molecular Weight:288.78 g/mol
- Merck Index:14,6488
- Melting Pt:290 °C (Zersetzung)
- Storage Temperature:Raumtemperatur
- EINECS:209-035-8
Dokumente
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



