TES, 0,2 M buffer soln., pH 7,5
TES buffer solution, 0,2 M, 7,5, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 70331-82-7
Physical Form: Liquid
MDL No.: MFCD00065482
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pk:250 mL
- Description:TES buffer solution
- pH value:7,5
- Cat. No.:J61350.AK
- Physical form:Liquid
- MDL No.:MFCD00065482
- CAS No.:70331-82-7



