Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, potassium phosphate, 0,2 M solution

Buffer, potassium phosphate, 0,2 M solution

Potassium phosphate, 0,2M buffer solution, pH 7,0, Glass bottle, 500 mL
Catalog # J61261.AP
Buffer, potassium phosphate, 0,2 M solution
Catalog # J61261.AP
CAS Number:  
Buffer, potassium phosphate, 0,2 M solution
Buffer, potassium phosphate, 0,2 M solution

Potassium phosphate, 0,2M buffer solution, pH 7,0, Glass bottle, 500 mL

CAS Number:  
Price
86,10 €
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Potassium phosphate, 0,2M buffer solution, pH 7,0
  • Cat. No.:
    J61261.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00011401
  • CAS No.:
    7778-77-0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.