Buffers, carbonate, 0,5 M solutions
Carbonate buffer solution, 0,5 M, 9,6, Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Buffers, carbonate, 0,5 M solutions
Catalog # J62610.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 497-19-8
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00003494
Documents
Specifications
Specifications
- Concentration:0,5 M
- Pack type:Glass bottle
- Pk:250 mL
- Description:Carbonate buffer solution
- pH value:9,6
- Cat. No.:J62610.AK
- Odour/scent:Odourless
- Physical form:Liquid
- MDL No.:MFCD00003494
- CAS No.:497-19-8



