Order Entry
Germany
Orders LinkContactUsLinkComponent
Kinase buffer I (5X)

Kinase buffer I (5X)

Kinase buffer I (5X), 250 mL
Catalog # J61967.AK
Kinase buffer I (5X)
Catalog # J61967.AK
CAS Number:  
Kinase buffer I (5X)
Kinase buffer I (5X)

Kinase buffer I (5X), 250 mL

CAS Number:  
Price
69,10 €
Log in for availability
1 * 250 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Kinase buffer I (5X)
  • Cat. No.:
    J61967.AK
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.