Order Entry
Germany
Orders LinkContactUsLinkComponent
Kinase buffer I (5X)

Kinase buffer I (5X)

Kinase buffer I (5X), 100 mL
Catalog # J61967.AE
Kinase buffer I (5X)
Catalog # J61967.AE
CAS Number:  
Kinase buffer I (5X)
Kinase buffer I (5X)

Kinase buffer I (5X), 100 mL

CAS Number:  
Price
47,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Kinase buffer I (5X)
  • Cat. No.:
    J61967.AE
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications