Order Entry
Germany
Orders LinkContactUsLinkComponent
 

LDS-sample buffer (4X), non-reducing

LDS-Sample Buffer (4X), non-reducing, 50 mL
Catalog # J61894.AD
LDS-sample buffer (4X), non-reducing
Catalog # J61894.AD
CAS Number:  
LDS-sample buffer (4X), non-reducing
LDS-sample buffer (4X), non-reducing

LDS-Sample Buffer (4X), non-reducing, 50 mL

CAS Number:  
Price
128,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.

Physical Form: Liquid
Appearance: Blue

This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    LDS-Sample Buffer (4X), non-reducing
  • Cat. No.:
    J61894.AD
  • Physical form:
    Liquid
  • Appearance:
    Blue

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.