Order Entry
Germany
Orders LinkContactUsLinkComponent
 

D(-)-Lactic acid

Glass bottle for solids, 1 g
Catalog # J61871.03
Corrosive
D(-)-Lactic acid
Catalog # J61871.03
CAS Number:  10326-41-7
D(-)-Lactic acid
D(-)-Lactic acid

Glass bottle for solids, 1 g

CAS Number:  10326-41-7
Price
906,00 €
Log in for availability
1 * 1 g
Product Details & Documents

CAS No.: 10326-41-7 Molecular Formula: C3H6O3 Formula Weight: 90.08 Storage Temperature: 2°C to 8°C Physical Form: Low melting solid Appearance: White to off-white Solubility: Soluble in water (50mg/ml) Beilstein Reference: 1720254 Merck Reference: 14,5335 Optical Rotation: -0.4° (c=8 in water) MDL No.: MFCD00068313

Useful chiral synthon, building block for depsipeptides

Synonyms:  (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
  • CAS Number:10326-41-7
  • Formula:C₃H₆O₃
  • MDL Number:MFCD00068313
  • Molecular Weight:90.08 g/mol
  • Merck Index:14,5335
  • Storage Temperature:Refrigerator
  • EINECS:233-713-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications