D(-)-Lactic acid
Glass bottle for solids, 250 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 10326-41-7 Molecular Formula: C3H6O3 Formula Weight: 90.08 Storage Temperature: 2°C to 8°C Physical Form: Low melting solid Appearance: White to off-white Solubility: Soluble in water (50mg/ml) Beilstein Reference: 1720254 Merck Reference: 14,5335 Optical Rotation: -0.4° (c=8 in water) MDL No.: MFCD00068313
Useful chiral synthon, building block for depsipeptides
Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
- CAS Number:10326-41-7
- Formula:C₃H₆O₃
- MDL Number:MFCD00068313
- Molecular Weight:90.08 g/mol
- Merck Index:14,5335
- Storage Temperature:Refrigerator
- EINECS:233-713-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



